AMBH324A4F0E Ambeed, Inc.

Dicyclohexylamine (S)-2-((tert-butoxycarbonyl)amino)-3-methoxypropanoate

  • MDL number MFCD09260875
  • Cas Number : 69912-63-6
  • Molecular Weight : 400.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/69912-63-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/69912-63-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart