AMBH324A4F13 Ambeed, Inc.

(1R,2R)-N1,N2-Dibenzylcyclohexane-1,2-diamine

  • MDL number MFCD29919494
  • Cas Number : 143443-23-6
  • Molecular Weight : 294.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/143443-23-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P310
MSDS https://www.ambeed.com/products/143443-23-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart