AMBH324A4F14 Ambeed, Inc.

(2R,3R,4R)-3,4-Bis(benzyloxy)-2-((benzyloxy)methyl)-5-methoxytetrahydrofuran

  • MDL number MFCD23704947
  • Cas Number : 64363-77-5
  • Molecular Weight : 434.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/64363-77-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/64363-77-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart