AMBH324A4F15 Ambeed, Inc.

trans-4'-(3,4-Difluorophenyl)-[1,1'-bi(cyclohexan)]-4-one

  • MDL number MFCD26143179
  • Cas Number : 147622-85-3
  • Molecular Weight : 292.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/147622-85-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/147622-85-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart