AMBH324A4F1A Ambeed, Inc.

Fmoc-allo-Thr-OH

  • MDL number MFCD01318739
  • Cas Number : 146306-75-4
  • Molecular Weight : 341.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/146306-75-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/146306-75-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart