AMBH324A4F1E Ambeed, Inc.

1,2,3-Trifluoro-5-[trans-4-[2-(trans-4-propylcyclohexyl)ethyl]cyclohexyl]benzene

  • MDL number MFCD12828865
  • Cas Number : 131819-24-4
  • Molecular Weight : 366.50

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/131819-24-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/131819-24-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart