AMBH324A4F23 Ambeed, Inc.

(E)-Ethyl 2-cyano-3-(4-hydroxyphenyl)acrylate

  • MDL number MFCD00188426
  • Cas Number : 6935-44-0
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/6935-44-0.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/6935-44-0.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart