AMBH324A4F2C Ambeed, Inc.

(-)-Corey Lactone Benzoate

  • MDL number MFCD00674102
  • Cas Number : 39746-00-4
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/39746-00-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/39746-00-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart