AMBH324A4F2E Ambeed, Inc.

sn-Glycero-3-phosphocholine

  • MDL number MFCD00063544
  • Cas Number : 28319-77-9
  • Molecular Weight : 257.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/28319-77-9.html

Safety Information

signalword Warning
pcodes P261-P273-P305+P351+P338
MSDS https://www.ambeed.com/products/28319-77-9.html
pictograms GHS07
hstatements H315-H319-H335-H412
Availability details for
Quantity
  • Availability
  • Add to Cart