AMBH324A4F34 Ambeed, Inc.

2-(2-(4-(3-(2-(Trifluoromethyl)-9H-thioxanthen-9-ylidene)propyl)piperazin-1-yl)ethoxy)ethanol dihydrochloride

  • Cas Number : 1535-17-7
  • Molecular Weight : 551.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1535-17-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1535-17-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart