AMBH324A4F40 Ambeed, Inc.

(S)-3,3-Dimethyltetrahydropyrrolo[1,2-c]oxazol-5(3H)-one

  • MDL number MFCD26940310
  • Cas Number : 99208-71-6
  • Molecular Weight : 155.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/99208-71-6.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/99208-71-6.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart