AMBH324A4F51 Ambeed, Inc.

(4S,4'S)-2,2'-(Propane-2,2-diyl)bis(4-(tert-butyl)-4,5-dihydrooxazole)

  • MDL number MFCD00192243
  • Cas Number : 131833-93-7
  • Molecular Weight : 294.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/131833-93-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/131833-93-7.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart