AMBH324A4F61 Ambeed, Inc.

H 89 Dihydrochloride

  • MDL number MFCD00214120
  • Cas Number : 130964-39-5
  • Molecular Weight : 519.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/130964-39-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/130964-39-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart