AMBH324A4F6E Ambeed, Inc.

(E)-1-Methoxy-4-(prop-1-en-1-yl)benzene

  • MDL number MFCD00009284
  • Cas Number : 4180-23-8
  • Molecular Weight : 148.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/4180-23-8.html

Safety Information

signalword Warning
pcodes P261-P280
MSDS https://www.ambeed.com/products/4180-23-8.html
pictograms GHS07
hstatements H317
Availability details for
Quantity
  • Availability
  • Add to Cart