AMBH324A4F70 Ambeed, Inc.

(S)-1-((S)-2-((tert-Butoxycarbonyl)amino)-3-methylbutanoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00076997
  • Cas Number : 23361-28-6
  • Molecular Weight : 314.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/23361-28-6.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/23361-28-6.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart