AMBH324A4F7B Ambeed, Inc.

H-Orn(Fmoc)-OH

  • MDL number MFCD00237208
  • Cas Number : 147071-84-9
  • Molecular Weight : 354.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/147071-84-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/147071-84-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart