AMBH324A4F92 Ambeed, Inc.

H-Ser(Bzl)-OMe.HCl

  • MDL number MFCD00191312
  • Cas Number : 19525-87-2
  • Molecular Weight : 245.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/19525-87-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/19525-87-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart