AMBH324A4F9E Ambeed, Inc.

(3R,7aR)-3-(tert-butyl)-1-hydroxy-7a-vinyltetrahydropyrrolo[1,2-c]oxazol-5(3H)-one

  • MDL number MFCD20040525
  • Cas Number : 1214741-21-5
  • Molecular Weight : 225.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1214741-21-5.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/1214741-21-5.html
pictograms GHS07
hstatements H302-H317
Availability details for
Quantity
  • Availability
  • Add to Cart