AMBH324A4FBF Ambeed, Inc.

2-(2-Aminothiazol-4-yl)-2-(methoxyimino)acetic acid

  • MDL number MFCD00071528
  • Cas Number : 65872-41-5
  • Molecular Weight : 201.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/65872-41-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/65872-41-5.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart