AMBH324A4FCA Ambeed, Inc.

H-Asp(Oet)-OEt.HCl

  • MDL number MFCD00038910
  • Cas Number : 16115-68-7
  • Molecular Weight : 225.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/16115-68-7.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/16115-68-7.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart