AMBH324A4FDC Ambeed, Inc.

1-(3,5-Bis(trifluoromethyl)phenyl)-3-((1R,2S)-2-hydroxy-2,3-dihydro-1H-inden-1-yl)thiourea

  • MDL number MFCD27922602
  • Cas Number : 871828-95-4
  • Molecular Weight : 420.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/871828-95-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/871828-95-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart