AMBH324A4FDD Ambeed, Inc.

(S)-1-((S)-3-mercapto-2-methylpropanoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00168073
  • Cas Number : 62571-86-2
  • Molecular Weight : 217.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/62571-86-2.html

Safety Information

signalword Danger
pcodes P280
MSDS https://www.ambeed.com/products/62571-86-2.html
pictograms GHS07,GHS08
hstatements H317-H361
Availability details for
Quantity
  • Availability
  • Add to Cart