AMBH324A4FED Ambeed, Inc.

H-Tyr(3,5-DiCl)-OH

  • MDL number MFCD09842574
  • Cas Number : 15106-62-4
  • Molecular Weight : 250.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/15106-62-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/15106-62-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart