AMBH324A4FF4 Ambeed, Inc.

Boc-Lys(Fmoc)-OH

  • MDL number MFCD00062032
  • Cas Number : 84624-27-1
  • Molecular Weight : 468.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/84624-27-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/84624-27-1.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart