AMBH324A4FF5 Ambeed, Inc.

(S)-1,5-Dimethylpiperazin-2-one hydrochloride

  • MDL number MFCD23096888
  • Cas Number : 1887197-43-4
  • Molecular Weight : 164.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1887197-43-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1887197-43-4.html
pictograms GHS07
hstatements H302-H315-H317-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart