AMBH324A4FFC Ambeed, Inc.

Ibutilide Fumarate

  • MDL number MFCD01715410
  • Cas Number : 122647-32-9
  • Molecular Weight : 885.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/122647-32-9.html

Safety Information

signalword Warning
pcodes P301+P312+P330
MSDS https://www.ambeed.com/products/122647-32-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart