AMBH324A4FFD Ambeed, Inc.

Boc-�HoAsp(OBzl)-OH

  • MDL number MFCD01862861
  • Cas Number : 254101-10-5
  • Molecular Weight : 337.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/254101-10-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/254101-10-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart