AMBH324A5002 Ambeed, Inc.

4'-((trans,trans)-4'-Ethyl-[1,1'-bi(cyclohexan)]-4-yl)-3,4,5-trifluoro-1,1'-biphenyl

  • MDL number MFCD12911826
  • Cas Number : 137529-40-9
  • Molecular Weight : 400.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/137529-40-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/137529-40-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart