AMBH324A501C Ambeed, Inc.

4'-(trans-4-propylcyclohexyl)-[1,1'-biphenyl]-4-carbonitrile

  • MDL number MFCD06658191
  • Cas Number : 94412-40-5
  • Molecular Weight : 303.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/94412-40-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/94412-40-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart