AMBH324A501D Ambeed, Inc.

4-(trans-4-Vinylcyclohexyl)benzonitrile

  • MDL number MFCD11053490
  • Cas Number : 96184-42-8
  • Molecular Weight : 211.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/96184-42-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/96184-42-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart