AMBH324A5030 Ambeed, Inc.

Tamsulosin

  • MDL number MFCD00871531
  • Cas Number : 106133-20-4
  • Molecular Weight : 408.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/106133-20-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/106133-20-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart