AMBH324A503A Ambeed, Inc.

Dichloro(cycloocta-1,5-diene)ruthenium(II)

  • MDL number MFCD00171304
  • Cas Number : 50982-12-2
  • Molecular Weight : 280.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/50982-12-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/50982-12-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart