AMBH324A503B Ambeed, Inc.

(S)-((2-Amino-4-(methylthio)butanoyl)oxy)zinc(II) hydrogensulfate

  • MDL number MFCD04118310
  • Cas Number : 56329-42-1
  • Molecular Weight : 310.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/56329-42-1.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/56329-42-1.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart