AMBH324A5041 Ambeed, Inc.

Boc-Arg(Pbf)-OH

  • MDL number MFCD00236821
  • Cas Number : 200124-22-7
  • Molecular Weight : 526.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/200124-22-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/200124-22-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart