AMBH324A5045 Ambeed, Inc.

(6S,8aR)-3-Oxooctahydroindolizine-6-carboxylic acid

  • MDL number MFCD28559196
  • Cas Number : 1620675-68-4
  • Molecular Weight : 183.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1620675-68-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1620675-68-4.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart