AMBH324A5046 Ambeed, Inc.

Boc-Hyp-OEt

  • MDL number MFCD09264314
  • Cas Number : 37813-30-2
  • Molecular Weight : 259.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/37813-30-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/37813-30-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart