AMBH324A506E Ambeed, Inc.

Boc-Cys(tBu)-OH

  • MDL number MFCD00076918
  • Cas Number : 56976-06-8
  • Molecular Weight : 277.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/56976-06-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/56976-06-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart