AMBH324A5081 Ambeed, Inc.

(S)-Di-tert-butyl 5-oxopyrrolidine-1,2-dicarboxylate

  • MDL number MFCD03094770
  • Cas Number : 91229-91-3
  • Molecular Weight : 285.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/91229-91-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/91229-91-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart