AMBH324A5087 Ambeed, Inc.

Boc-Lysinol(Z)

  • MDL number MFCD00235934
  • Cas Number : 82689-20-1
  • Molecular Weight : 366.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/82689-20-1.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/82689-20-1.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart