AMBH324A508F Ambeed, Inc.

Mibefradil dihydrochloride

  • MDL number MFCD00881804
  • Cas Number : 116666-63-8
  • Molecular Weight : 568.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/116666-63-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/116666-63-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart