AMBH324A5090 Ambeed, Inc.

(R)-2-Hydroxy-N-(4-nitrophenethyl)-2-phenylacetamide

  • MDL number MFCD22200231
  • Cas Number : 521284-19-5
  • Molecular Weight : 300.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/521284-19-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/521284-19-5.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart