AMBH324A509A Ambeed, Inc.

(R)-2-Chloro-7-ethyl-8-isopropyl-5-methyl-7,8-dihydropteridin-6(5H)-one

  • MDL number MFCD16621126
  • Cas Number : 877676-50-1
  • Molecular Weight : 268.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/877676-50-1.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/877676-50-1.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart