AMBH324A509B Ambeed, Inc.

(S)-2-Hydroxybutanoic acid

  • MDL number MFCD01318568
  • Cas Number : 3347-90-8
  • Molecular Weight : 104.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/3347-90-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/3347-90-8.html
pictograms GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart