AMBH324A509F Ambeed, Inc.

Diethyl Azodicarboxylate (DEAD)

  • MDL number MFCD00009103
  • Cas Number : 1972-28-7
  • Molecular Weight : 174.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/1972-28-7.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1972-28-7.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart