AMBH324A50A5 Ambeed, Inc.

(R)-Ethyl 2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate

  • MDL number MFCD18071557
  • Cas Number : 1146954-88-2
  • Molecular Weight : 233.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1146954-88-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/1146954-88-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart