AMBH324A50AC Ambeed, Inc.

(2S,3S)-2-((2S,3S)-2-(((Benzyloxy)carbonyl)amino)-3-methylpentanamido)-3-methylpentanoic acid

  • MDL number MFCD00027066
  • Cas Number : 42538-01-2
  • Molecular Weight : 378.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/42538-01-2.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/42538-01-2.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart