AMBH324A50B8 Ambeed, Inc.

4-Fluoro-4'-(trans-4-propylcyclohexyl)-1,1'-biphenyl

  • MDL number MFCD11053438
  • Cas Number : 87260-24-0
  • Molecular Weight : 296.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/87260-24-0.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/87260-24-0.html
pictograms GHS07
hstatements H315-H319-H413
Availability details for
Quantity
  • Availability
  • Add to Cart