AMBH324A50BA Ambeed, Inc.

Meropenem trihydrate

  • MDL number MFCD08600005
  • Cas Number : 119478-56-7
  • Molecular Weight : 437.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/119478-56-7.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/119478-56-7.html
pictograms GHS07
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart