AMBH324A50BE Ambeed, Inc.

H-Tyr(tBu)-OtBu.HCl

  • MDL number MFCD00058002
  • Cas Number : 17083-23-7
  • Molecular Weight : 329.86

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/17083-23-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/17083-23-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart