AMBH324A50C2 Ambeed, Inc.

Ceftibuten dihydrate

  • MDL number MFCD08141796
  • Cas Number : 118081-34-8
  • Molecular Weight : 446.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/118081-34-8.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/118081-34-8.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart